Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20102G100EA 0 CHF
ACTIAS20102G100
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel: C₁₉H₁₅NO₅
MW: 337,3 g/mol
MDL Number: MFCD00010733
CAS-Nummer: 82911-69-1
EINECS: 433-520-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR