Ethylcyclobutancarboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS24310G100 AS24310G25
ACTIAS24310G100EA 106 CHF
ACTIAS24310G100 ACTIAS24310G25
Ethylcyclobutancarboxylat
Ethylcyclobutancarboxylat
Formel: C₇H₁₂O₂
MW: 128,1690 g/mol
MDL Number: MFCD00001321
CAS-Nummer: 14924-53-9
EINECS: 238-995-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR