2-Aminopyrimidin-5-carbosäurenitril

Lieferant: ACTIVATE SCIENTIFIC

AS30195G1 AS30195G5
ACTIAS30195G1EA 0 CHF
ACTIAS30195G1 ACTIAS30195G5
2-Aminopyrimidin-5-carbosäurenitril
2-Aminopyrimidin-5-carbosäurenitril
Formel: C₅H₄N₄
MDL Number: MFCD00232791
CAS-Nummer: 1753-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR