4-Amino-2-brom-5-pyrimidincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS34960G1 AS34960G5
ACTIAS34960G1EA 0 CHF
ACTIAS34960G1 ACTIAS34960G5
4-Amino-2-brom-5-pyrimidincarbonitril
4-Amino-2-brom-5-pyrimidincarbonitril
Formel: C₅H₃BrN₄
MW: 199,01 g/mol
MDL Number: MFCD00173661
CAS-Nummer: 94741-70-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR