2-Methyltryptamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(2-Aminoethyl)-2-methylindole

AS31025G1 AS31025G5
ACTIAS31025G1EA 0 CHF
ACTIAS31025G1 ACTIAS31025G5
2-Methyltryptamin
2-Methyltryptamin
Formel: C₁₁H₁₄N₂
MDL Number: MFCD00130185
CAS-Nummer: 2731-06-8
EINECS: 220-347-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR