2-Amino-3-methylbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-6-(trifluoromethyl)aniline

AS32498G1 AS32498G5
ACTIAS32498G1EA 0 CHF
ACTIAS32498G1 ACTIAS32498G5
2-Amino-3-methylbenzotrifluorid
2-Amino-3-methylbenzotrifluorid
Formel: C₈H₈F₃N
MW: 175,1500 g/mol
MDL Number: MFCD03701069
CAS-Nummer: 88301-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR