3-Amino-4-methylbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-5-(trifluoromethyl)aniline

AS34935G1 AS34935G10 AS34935G5
ACTIAS34935G1EA 0 CHF
ACTIAS34935G1 ACTIAS34935G10 ACTIAS34935G5
3-Amino-4-methylbenzotrifluorid
3-Amino-4-methylbenzotrifluoride
Formel: C₈H₈F₃N
MW: 175,16 g/mol
MDL Number: MFCD00134580
CAS-Nummer: 25449-96-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR