4-Amino-3-methylbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-4-(trifluoromethyl)aniline

AS35801G1 AS35801G5
ACTIAS35801G1EA 0 CHF
ACTIAS35801G1 ACTIAS35801G5
4-Amino-3-methylbenzotrifluorid
4-Amino-3-methylbenzotrifluorid
Formel: C₈H₈F₃N
MW: 175,15 g/mol
MDL Number: MFCD03701068
CAS-Nummer: 67169-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR