2,6-Dichlornicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dichloropyridine-3-carbonitrile, 2,6-Dichloro-3-cyanopyridine

AS22396G1 AS22396G5
ACTIAS22396G1EA 0 CHF
ACTIAS22396G1 ACTIAS22396G5
2,6-Dichlornicotinonitril
2,6-Dichlornicotinonitril
Formel: C₆H₂Cl₂N₂
MW: 173,00 g/mol
MDL Number: MFCD03411341
CAS-Nummer: 40381-90-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR