5-Brom-2-methoxy-6-methylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine
AS33569G1
AS33569G10
AS33569G5
ACTIAS33569G1EA
0
CHF
ACTIAS33569G1
ACTIAS33569G10
ACTIAS33569G5
5-Brom-2-methoxy-6-methylpyridin
5-Brom-2-methoxy-6-methylpyridin
Formel:
C₇H₈BrNO MW: 202,05 g/mol |
MDL Number:
MFCD03094943 CAS-Nummer: 126717-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...