4-Chlor-8-fluorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS35896G1 AS35896G5
ACTIAS35896G1EA 0 CHF
ACTIAS35896G1 ACTIAS35896G5
4-Chlor-8-fluorchinolin
4-Chloro-8-fluoroquinoline
Formel: C₉H₅ClFN
MDL Number: MFCD00278785
CAS-Nummer: 63010-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR