(3-Methoxy-4-methylphenyl)acetonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32446G1 AS32446G5
ACTIAS32446G1EA 0 CHF
ACTIAS32446G1 ACTIAS32446G5
(3-Methoxy-4-methylphenyl)acetonitril
(3-Methoxy-4-methylphenyl)acetonitril
Formel: C₁₀H₁₁NO
MDL Number: MFCD00060309
CAS-Nummer: 64829-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR