Methyl-2-amino-5-brombenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-amino-5-bromobenzoate

AS35912G1 AS35912G5
ACTIAS35912G1EA 0 CHF
ACTIAS35912G1 ACTIAS35912G5
Methyl-2-amino-5-brombenzoat
Methyl-2-amino-5-brombenzoat
Formel: C₈H₈BrNO₂
MDL Number: MFCD00144761
CAS-Nummer: 52727-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR