4-Brom-3-fluorphenylessigsäure

Lieferant: ACTIVATE SCIENTIFIC

AS32519G1 AS32519G5
ACTIAS32519G1EA 37.48 CHF
ACTIAS32519G1 ACTIAS32519G5
4-Brom-3-fluorphenylessigsäure
4-Brom-3-fluorphenylessigsäure
Formel: C₈H₆BrFO₂
MW: 233,0300 g/mol
MDL Number: MFCD12756200
CAS-Nummer: 942282-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR