4-Brom-2-fluorphenylessigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(4-Bromo-2-fluorophenyl)acetic acid

AS33568G1 AS33568G5
ACTIAS33568G1EA 0 CHF
ACTIAS33568G1 ACTIAS33568G5
4-Brom-2-fluorphenylessigsäure
4-Brom-2-fluorphenylessigsäure
Formel: C₈H₆BrFO₂
MW: 233,03 g/mol
MDL Number: MFCD09032952
CAS-Nummer: 114897-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR