2-Brom-4-methylthiazol

Lieferant: ACTIVATE SCIENTIFIC

AS50056G1 AS50056G5
ACTIAS50056G1EA 0 CHF
ACTIAS50056G1 ACTIAS50056G5
2-Brom-4-methylthiazol
2-Bromo-4-methylthiazole
Formel: C₄H₄BrNS
MDL Number: MFCD06660130
CAS-Nummer: 7238-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR