5-Thiazolcarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
AS50101G1
AS50101G5
ACTIAS50101G1EA
32.28
CHF
ACTIAS50101G1
ACTIAS50101G5
5-Thiazolcarbonsäure
5-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S MW: 129,1371 g/mol |
MDL Number:
MFCD03428539 CAS-Nummer: 14527-41-4 EINECS: 238-545-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...