4-Thiazolcarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Thiazole-4-carboxylic acid
AS50100G100
AS50100G5
ACTIAS50100G100EA
0
CHF
ACTIAS50100G100
ACTIAS50100G5
4-Thiazolcarbonsäure
4-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S MW: 129,1371 g/mol |
MDL Number:
MFCD00623589 CAS-Nummer: 3973-08-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...