2-Thiazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Thiazole-2-carboxylic acid

AS30366G1 AS30366G5
ACTIAS30366G1EA 0 CHF
ACTIAS30366G1 ACTIAS30366G5
2-Thiazolcarbonsäure
2-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
MW: 129,14 g/mol
MDL Number: MFCD02094169
CAS-Nummer: 14190-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR