2-Brom-5-chlorthiazol

Lieferant: ACTIVATE SCIENTIFIC

AS31938G1 AS31938G5
ACTIAS31938G1EA 0 CHF
ACTIAS31938G1 ACTIAS31938G5
2-Brom-5-chlorthiazol
2-Bromo-5-chlorothiazole
Formel: C₃HBrClNS
MW: 198,4687 g/mol
MDL Number: MFCD09909626
CAS-Nummer: 16629-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR