2-Brom-5-formylthiazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromothiazole-5-carboxaldehyde

AS50070G1 AS50070G5
ACTIAS50070G1EA 0 CHF
ACTIAS50070G1 ACTIAS50070G5
2-Brom-5-formylthiazol
2-Brom-5-formylthiazol
Formel: C₄H₂BrNOS
MW: 192.03 g/mol
MDL Number: MFCD03788567
CAS-Nummer: 464192-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR