(±)-3-Aminochinuclidin Dihydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

AS21454G25 AS21454G5
ACTIAS21454G25EA 241 CHF
ACTIAS21454G25 ACTIAS21454G5
(±)-3-Aminochinuclidin Dihydrochlorid
(±)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
MW: 199,1213 g/mol
MDL Number: MFCD00137395
CAS-Nummer: 6530-09-2
EINECS: 229-424-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR