(S)-(-)-3-Aminochinuclidin Dihydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

AS35003G1 AS35003G10 AS35003G5
ACTIAS35003G1EA 0 CHF
ACTIAS35003G1 ACTIAS35003G10 ACTIAS35003G5
(S)-(-)-3-Aminochinuclidin Dihydrochlorid
(S)-(-)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
MW: 199,1213 g/mol
MDL Number: MFCD00191753
CAS-Nummer: 119904-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR