5-Formylfuran-2-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Formylfuran-5-boronic acid, (5-formylfuran-2-yl)boronic acid, 5-formylfuran-2-boronic acid, (5-Formylfuran-2-yl)boronic acid, 5-Formyl-2-furanboronic acid

AS51433G1 AS51433G5
ACTIAS51433G1EA 0 CHF
ACTIAS51433G1 ACTIAS51433G5
5-Formylfuran-2-boronsäure
5-Formylfuran-2-boronsäure
Formel: C₅H₅BO₄
MW: 139.90 g/mol
MDL Number: MFCD01114696
CAS-Nummer: 27329-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR