4-Brom-1,3-benzodioxol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromobenzo[d][1,3]dioxole

AS36643G1 AS36643G5
ACTIAS36643G1EA 0 CHF
ACTIAS36643G1 ACTIAS36643G5
4-Brom-1,3-benzodioxol
4-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
MW: 201,0174 g/mol
MDL Number: MFCD02681890
CAS-Nummer: 6698-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR