3,5-Dibrom-4-picolin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3,5-Dibromo-4-methylpyridine
AS31252G1
AS31252G5
ACTIAS31252G1EA
0
CHF
ACTIAS31252G1
ACTIAS31252G5
3,5-Dibrom-4-picolin
3,5-Dibrom-4-picolin
Formel:
C₆H₅Br₂N MW: 250.92 g/mol |
MDL Number:
MFCD00128859 CAS-Nummer: 3430-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...