4-Brom-2,6-dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS36358G1 AS36358G5
ACTIAS36358G1EA 0 CHF
ACTIAS36358G1 ACTIAS36358G5
4-Brom-2,6-dichlorpyridin
4-Bromo-2,6-dichloropyridine
Formel: C₅H₂BrCl₂N
MDL Number: MFCD11848646
CAS-Nummer: 98027-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR