2-Brom-5-methylbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

AS33762G1 AS33762G10 AS33762G5
ACTIAS33762G1EA 0 CHF
ACTIAS33762G1 ACTIAS33762G10 ACTIAS33762G5
2-Brom-5-methylbenzotrifluorid
2-Brom-5-methylbenzotrifluorid
Formel: C₈H₆BrF₃
MDL Number: MFCD01631586
CAS-Nummer: 261952-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR