4-Fluor-2-nitrobenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-2-nitro-1-(trifluoromethyl)benzene

AS33412G1 AS33412G5
ACTIAS33412G1EA 0 CHF
ACTIAS33412G1 ACTIAS33412G5
4-Fluor-2-nitrobenzotrifluorid
4-Fluor-2-nitrobenzotrifluorid
Formel: C₇H₃F₄NO₂
CAS-Nummer: 182289-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR