2-Fluor-3-nitrobenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

AS32672G1 AS32672G5
ACTIAS32672G1EA 0 CHF
ACTIAS32672G1 ACTIAS32672G5
2-Fluor-3-nitrobenzotrifluorid
2-Fluor-3-nitrobenzotrifluorid
Formel: C₇H₃F₄NO₂
MW: 209.10 g/mol
MDL Number: MFCD08532487
CAS-Nummer: 61324-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR