3-Chlor-5-nitrobenzotrifluorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Chloro-3-nitro-5-(trifluoromethyl)benzene
AS33808G1
AS33808G10
AS33808G5
ACTIAS33808G1EA
0
CHF
ACTIAS33808G1
ACTIAS33808G10
ACTIAS33808G5
3-Chlor-5-nitrobenzotrifluorid
3-Chlor-5-nitrobenzotrifluorid
Formel:
C₇H₃ClF₃NO₂ MW: 225,5500 g/mol |
MDL Number:
MFCD03412202 CAS-Nummer: 401-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...