2-Brom-3,5-dichloranilin

Lieferant: ACTIVATE SCIENTIFIC

AS33937G1 AS33937G5
ACTIAS33937G1EA 0 CHF
ACTIAS33937G1 ACTIAS33937G5
2-Brom-3,5-dichloranilin
2-Brom-3,5-dichloranilin
Formel: C₆H₄BrCl₂N
MDL Number: MFCD09999825
CAS-Nummer: 1211214-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR