Methyl-2-amino-5-methoxybenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-amino-5-methoxybenzoate

AS31670G1 AS31670G5
ACTIAS31670G1EA 0 CHF
ACTIAS31670G1 ACTIAS31670G5
Methyl-2-amino-5-methoxybenzoat
Methyl-2-amino-5-methoxybenzoat
Formel: C₉H₁₁NO₃
MW: 181,19 g/mol
MDL Number: MFCD09038482
CAS-Nummer: 2475-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR