2,6-Difluor-4-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS30796G1 AS30796G5
ACTIAS30796G1EA 0 CHF
ACTIAS30796G1 ACTIAS30796G5
2,6-Difluor-4-nitrophenol
2,6-Difluor-4-nitrophenol
Formel: C₆H₃F₂NO₃
MW: 175.0900 g/mol
MDL Number: MFCD07366521
CAS-Nummer: 658-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR