(R)-(+)-β-Methylphenethylamin

Lieferant: ACTIVATE SCIENTIFIC

AS33136G1 AS33136G5
ACTIAS33136G1EA 0 CHF
ACTIAS33136G1 ACTIAS33136G5
(R)-(+)-β-Methylphenethylamin
(R)-(+)-β-Methylphenethylamin
Formel: C₉H₁₃N
MDL Number: MFCD00216740
CAS-Nummer: 28163-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR