1,3-Benzodioxol-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Benzo[d][1,3]dioxole-4-carboxylic acid

AS20846G1 AS20846G5
ACTIAS20846G1EA 98 CHF
ACTIAS20846G1 ACTIAS20846G5
1,3-Benzodioxol-4-carbonsäure
1,3-Benzodioxol-4-carbonsäure
Formel: C₈H₆O₄
MW: 166,1308 g/mol
MDL Number: MFCD01076411
CAS-Nummer: 5768-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR