4-Chlor-3-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS16231G1 AS16231G5
ACTIAS16231G1EA 0 CHF
ACTIAS16231G1 ACTIAS16231G5
4-Chlor-3-iodpyridin
4-Chloro-3-iodopyridine
Formel: C₅H₃ClIN
MW: 239,4415 g/mol
MDL Number: MFCD00234024
CAS-Nummer: 89167-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR