2,6-Dichlor-4-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS33373G1
AS33373G5
ACTIAS33373G1EA
0
CHF
ACTIAS33373G1
ACTIAS33373G5
2,6-Dichlor-4-iodpyridin
2,6-Dichloro-4-iodopyridine
Formel:
C₅H₂CI₂IN MW: 273,89 g/mol |
MDL Number:
MFCD07368400 CAS-Nummer: 98027-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...