2,3-Dichlor-4-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35359G1 AS35359G5
ACTIAS35359G1EA 0 CHF
ACTIAS35359G1 ACTIAS35359G5
2,3-Dichlor-4-iodpyridin
2,3-Dichloro-4-iodopyridine
Formel: C₅H₂Cl₂IN
MW: 273,8900 g/mol
MDL Number: MFCD11857737
CAS-Nummer: 889865-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR