2-(2,2,2-Trifluoracetyl)thiophen

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(Trifluoroacetyl)thiophene

AS20657G25 AS20657G5
ACTIAS20657G25EA 727 CHF
ACTIAS20657G25 ACTIAS20657G5
2-(2,2,2-Trifluoracetyl)thiophen
2-(2,2,2-Trifluoracetyl)thiophen
Formel: C₆H₅ClOS
MW: 180,15 g/mol
MDL Number: MFCD00041427
CAS-Nummer: 651-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR