2-Iod-3-pyridinol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Hydroxy-2-iodopyridine
AS31280G1
AS31280G10
AS31280G5
ACTIAS31280G1EA
0
CHF
ACTIAS31280G1
ACTIAS31280G10
ACTIAS31280G5
2-Iod-3-pyridinol
2-Iod-3-pyridinol
Formel:
C₅H₄INO MW: 221,00 g/mol |
MDL Number:
MFCD00023421 CAS-Nummer: 40263-57-8 EINECS: 254-864-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...