4-(Dimethylamino)cyclohexanon

Lieferant: ACTIVATE SCIENTIFIC

AS34188G1 AS34188G5
ACTIAS34188G1EA 0 CHF
ACTIAS34188G1 ACTIAS34188G5
4-(Dimethylamino)cyclohexanon
4-(Dimethylamino)cyclohexanone
Formel: C₈H₁₅NO
MW: 141.2100 g/mol
MDL Number: MFCD09037892
CAS-Nummer: 40594-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR