4-Oxocyclohexancarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Oxo-cyclohexanecarbonitrile

AS23420G1 AS23420G5
ACTIAS23420G1EA 37 CHF
ACTIAS23420G1 ACTIAS23420G5
4-Oxocyclohexancarbonitril
4-Oxocyclohexancarbonitril
Formel: C₇H₉NO
MDL Number: MFCD11847831
CAS-Nummer: 34916-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR