O-Benzylhydroxylamin Hydrochlorid
Lieferant: ACTIVATE SCIENTIFIC
AS21992G100
AS21992G25
ACTIAS21992G100EA
0
CHF
ACTIAS21992G100
ACTIAS21992G25
O-Benzylhydroxylamin Hydrochlorid
O-Benzylhydroxylamin Hydrochlorid
Formel:
C₇H₁₀ClNO |
MDL Number:
MFCD00012952 CAS-Nummer: 2687-43-6 EINECS: 220-249-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...