4-Brom-2-pyridon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

AS30593G1 AS30593G5
ACTIAS30593G1EA 0 CHF
ACTIAS30593G1 ACTIAS30593G5
4-Brom-2-pyridon
4-Brom-2-pyridon
Formel: C₅H₄BrNO
MW: 174,00 g/mol
MDL Number: MFCD00234041
CAS-Nummer: 36953-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR