3-Brom-4-pyridon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-4-hydroxypyridine, 3-Bromo-4-pyridinol

AS30279G1 AS30279G5
ACTIAS30279G1EA 0 CHF
ACTIAS30279G1 ACTIAS30279G5
3-Brom-4-pyridon
3-Brom-4-pyridon
Formel: C₅H₄BrNO
MW: 174,00 g/mol
MDL Number: MFCD01646107
CAS-Nummer: 36953-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR