3,5-Dibrom-4-pyridon

Lieferant: ACTIVATE SCIENTIFIC

AS30336G1 AS30336G5
ACTIAS30336G1EA 0 CHF
ACTIAS30336G1 ACTIAS30336G5
3,5-Dibrom-4-pyridon
3,5-Dibrom-4-pyridon
Formel: C₅H₃Br₂NO
MDL Number: MFCD00082969
CAS-Nummer: 141375-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR