2,6-Dibrom-3-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dibromopyridin-3-ol

AS36016G1 AS36016G5
ACTIAS36016G1EA 0 CHF
ACTIAS36016G1 ACTIAS36016G5
2,6-Dibrom-3-hydroxypyridin
2,6-Dibromo-3-hydroxypyridine
Formel: C₅H₃Br₂NO
MW: 252.8900 g/mol
MDL Number: MFCD01318479
CAS-Nummer: 6602-33-1
EINECS: 229-548-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR