2-Amino-3-bromphenol

Lieferant: ACTIVATE SCIENTIFIC

AS32236G1 AS32236G5
ACTIAS32236G1EA 0 CHF
ACTIAS32236G1 ACTIAS32236G5
2-Amino-3-bromphenol
2-Amino-3-bromphenol
Formel: C₆H₆BrNO
MW: 188,0200 g/mol
MDL Number: MFCD08445676
CAS-Nummer: 116435-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR