2-Chlorresorcin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chlorobenzene-1,3-diol

AS30383G10 AS30383G5
ACTIAS30383G10EA 0 CHF
ACTIAS30383G10 ACTIAS30383G5
2-Chlorresorcin
2-Chlorresorcin
Formel: C₆H₅ClO₂
MDL Number: MFCD06658175
CAS-Nummer: 6201-65-6
EINECS: 228-256-0

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR